1-(Piperidin-4-yl)-2-(trifluoromethyl)-1H-benzo[d]imidazole ≥95%

Supplier: Matrix Scientific

072405-250MG 072405-1G
102524-676EA 640 USD
102524-676 102524-678
1-(Piperidin-4-yl)-2-(trifluoromethyl)-1H-benzo[d]imidazole ≥95%
1-(Piperidin-4-yl)-2-(trifluoromethyl)-1H-benzo[d]imidazole

Hazard: Irritant

Formula: C₁₃H₁₄F₃N₃
MW: 269.27 g/mol
MDL Number: MFCD09999148
CAS Number: 742076-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR