2-(Methylamino)pyrimidine-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

072404-250MG 072404-1G 072404-5G
102524-670EA 640 USD
102524-670 102524-672 102524-674
2-(Methylamino)pyrimidine-4-carbaldehyde ≥95%
2-(Methylamino)pyrimidine-4-carbaldehyde

Hazard: Irritant

Formula: C₆H₇N₃O
CAS Number: 180869-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR