tert-Butyl-5-methoxy-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 5-methoxy-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate

072403-250MG 072403-1G
102524-666EA 1223.08 USD
102524-666 102524-668
tert-Butyl-5-methoxy-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate ≥95%
tert-Butyl-5-methoxy-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate

Hazard: Irritant

Formula: C₁₈H₂₄N₂O₄
MW: 332.3942 g/mol
MDL Number: MFCD10566055
CAS Number: 752234-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR