tert-Butyl-5-chloro-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 5-chloro-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate

072402-250MG 072402-1G
102524-662EA 1223.08 USD
102524-662 102524-664
tert-Butyl-5-chloro-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate ≥95%
tert-Butyl-5-chloro-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate

Hazard: Irritant

Formula: C₁₇H₂₁ClN₂O₃
MW: 336.82 g/mol
MDL Number: MFCD10566053
CAS Number: 332187-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR