tert-Butyl-5-bromo-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 5-bromo-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate

072401-250MG 072401-1G
102524-658EA 785.92 USD
102524-658 102524-660
tert-Butyl-5-bromo-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate ≥95%
tert-Butyl-5-bromo-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate

Hazard: Irritant

Formula: C₁₇H₂₁BrN₂O₃
MW: 381.2600 g/mol
MDL Number: MFCD10566052
CAS Number: 873779-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR