1'-Boc-1,2-dihydro-2-oxospiro[3H-indole-3,4'-piperidine] ≥95%

Supplier: Matrix Scientific

Synonyms: Spiro[indole-3,4'-piperidin]-2(1H)-one, N1'-BOC protected, 1'-Boc-1,2-dihydro-2-oxo-spiro[3H-indole-3,4'-piperidine]

072400-250MG 072400-1G 072400-5G
102524-652EA 481.82 USD
102524-652 102524-654 102524-656
1'-Boc-1,2-dihydro-2-oxospiro[3H-indole-3,4'-piperidine] ≥95%
1'-Boc-1,2-dihydro-2-oxospiro[3H-indole-3,4'-piperidine]

Hazard: Irritant

Formula: C₁₇H₂₂N₂O₃
MW: 302.3682 g/mol
MDL Number: MFCD10566051
CAS Number: 252882-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR