1'-Methylspiro[indoline-3,4'-piperidine] ≥95%

Supplier: Matrix Scientific

072399-250MG 072399-1G
102524-648EA 1223.08 USD
102524-648 102524-650
1'-Methylspiro[indoline-3,4'-piperidine] ≥95%
1'-Methylspiro[indoline-3,4'-piperidine]

Hazard: Irritant

Formula: C₁₃H₁₈N₂
MW: 202.2954 g/mol
MDL Number: MFCD12198567
CAS Number: 69584-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR