6-Methoxy-3-methyl-1H-indazole ≥95%

Supplier: Matrix Scientific

072398-250MG 072398-1G 072398-5G
102524-642EA 498.46 USD
102524-642 102524-644 102524-646
6-Methoxy-3-methyl-1H-indazole ≥95%
6-Methoxy-3-methyl-1H-indazole

Hazard: Irritant

Formula: C₉H₁₀N₂O
MDL Number: MFCD07780793
CAS Number: 7746-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR