5-Bromo-3-methyl-1H-indazole ≥95%

Supplier: Matrix Scientific

072396-1G 072396-5G
102524-632EA 250.77 USD
102524-632 102524-634
5-Bromo-3-methyl-1H-indazole ≥95%
5-Bromo-3-methyl-1H-indazole

Hazard: Irritant

Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
MDL Number: MFCD05664003
CAS Number: 552331-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR