6-Hydroxy-3-methylindazole ≥95%

Supplier: Matrix Scientific

072394-250MG 072394-1G 072394-5G
102524-618EA 498.46 USD
102524-618 102524-620 102524-622
6-Hydroxy-3-methylindazole ≥95%
6-Hydroxy-3-methylindazole

Hazard: Irritant

Formula: C₈H₈N₂O
MW: 148.1600 g/mol
MDL Number: MFCD08692367
CAS Number: 201286-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR