5-Bromobenzo[d]isoxazole ≥95%

Supplier: Matrix Scientific

072393-250MG 072393-1G 072393-5G
102524-612EA 400 USD
102524-612 102524-614 102524-616
5-Bromobenzo[d]isoxazole ≥95%
5-Bromobenzo[d]isoxazole

Hazard: Irritant

Formula: C₇H₄BrNO
MW: 198.0200 g/mol
MDL Number: MFCD06659644
CAS Number: 837392-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR