3-Ethynylpyrazin-2-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Ethynyl-2-pyrazinamine, 2-Amino-3-ethynylpyrazine

072392-250MG 072392-1G
102524-608EA 924.62 USD
102524-608 102524-610
3-Ethynylpyrazin-2-amine ≥95%
3-Ethynylpyrazin-2-amine

Hazard: Irritant

Formula: C6H5N3
MW: 119.1240 g/mol
MDL Number: MFCD09999143
CAS Number: 1005349-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR