2-Amino-5-bromo-3-ethynylpyrazine ≥95%
Supplier: Matrix Scientific
072391-250MG
072391-1G
072391-5G
102524-602EA
498.46
USD
102524-602
102524-604
102524-606
2-Amino-5-bromo-3-ethynylpyrazine ≥95%
2-Amino-5-bromo-3-ethynylpyrazine
Hazard: Irritant
Formula:
C₆H₄BrN₃ MW: 198.0201 g/mol |
MDL Number:
MFCD09999142 CAS Number: 1209289-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...