4-Amino-2,6-difluorophenol hydrochloride ≥95%

Supplier: Matrix Scientific

072387-250MG 072387-1G 072387-5G
102524-580EA 250.77 USD
102524-580 102524-582 102524-584
4-Amino-2,6-difluorophenol hydrochloride ≥95%
4-Amino-2,6-difluorophenol hydrochloride

Hazard: Irritant

Formula: C₆H₆ClF₂NO
MDL Number: MFCD12197391
CAS Number: 220353-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR