2,6-Difluoro-4-nitrophenol ≥95%

Supplier: Matrix Scientific

072385-1G 072385-5G 072385-10G
102524-568EA 533.85 USD
102524-568 102524-570 102524-572
2,6-Difluoro-4-nitrophenol ≥95%
2,6-Difluoro-4-nitrophenol

Hazard: Irritant

Formula: C₆H₃F₂NO₃
MDL Number: MFCD07366521
CAS Number: 658-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR