1H-Pyrazol-4-amine hydrochloride ≥95%

Supplier: Matrix Scientific

072384-1G 072384-5G 072384-10G
102524-562EA 533.85 USD
102524-562 102524-564 102524-566
1H-Pyrazol-4-amine hydrochloride ≥95%
1H-Pyrazol-4-amine hydrochloride

Hazard: Irritant

Formula: C₃H₆ClN₃
MW: 119.55 g/mol
MDL Number: MFCD01693708
CAS Number: 4331-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR