6-Fluorobenzo[d]isoxazol-3-ylamine ≥95%

Supplier: Matrix Scientific

072383-250MG 072383-1G 072383-5G
102524-556EA 423.69 USD
102524-556 102524-558 102524-560
6-Fluorobenzo[d]isoxazol-3-ylamine ≥95%
6-Fluorobenzo[d]isoxazol-3-ylamine

Hazard: Irritant

Formula: C₇H₅FN₂O
MW: 152.1300 g/mol
MDL Number: MFCD09999138
CAS Number: 177995-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR