5-Fluorobenzo[d]isoxazol-3-amine ≥95%

Supplier: Matrix Scientific

072382-1G 072382-5G
102524-552EA 323.08 USD
102524-552 102524-554
5-Fluorobenzo[d]isoxazol-3-amine ≥95%
5-Fluorobenzo[d]isoxazol-3-amine

Hazard: Irritant

Formula: C7H5FN2O
MW: 152.1300 g/mol
MDL Number: MFCD09999137
CAS Number: 868271-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR