2-Bromo-5-hydroxybenzonitrile ≥95%

Supplier: Matrix Scientific

072381-1G 072381-5G
102524-546EA 290.77 USD
102524-546 102524-548
2-Bromo-5-hydroxybenzonitrile ≥95%
2-Bromo-5-hydroxybenzonitrile

Hazard: Irritant

Formula: C₇H₄BrNO
MW: 198.0200 g/mol
MDL Number: MFCD08061922
CAS Number: 189680-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR