6-Methoxybenzo[d]thiazol-2(3H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Methoxy-2(3H)-benzothiazolone

072378-250MG 072378-1G 072378-5G
102524-528EA 355.38 USD
102524-528 102524-530 102524-532
6-Methoxybenzo[d]thiazol-2(3H)-one ≥95%
6-Methoxybenzo[d]thiazol-2(3H)-one

Hazard: Irritant

Formula: C₈H₇NO₂S
MDL Number: MFCD09743851
CAS Number: 40925-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR