1,4-Dibromoisoquinoline ≥95%

Supplier: Matrix Scientific

072377-250MG 072377-1G
102524-524EA 581.43 USD
102524-524 102524-526
1,4-Dibromoisoquinoline ≥95%
1,4-Dibromoisoquinoline

Hazard: Irritant

Formula: C₉H₅Br₂N
MW: 286.9507 g/mol
MDL Number: MFCD00234475
CAS Number: 51206-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR