2-Amino-4-bromobenzaldehyde ≥95%

Supplier: Matrix Scientific

072375-1G 072375-5G
102524-512EA 106.15 USD
102524-512 102524-514
2-Amino-4-bromobenzaldehyde ≥95%
2-Amino-4-bromobenzaldehyde

Hazard: Irritant

Formula: C₇H₆BrNO
Melting Pt: 85 °C
MDL Number: MFCD08458822
CAS Number: 59278-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR