1-(5-Methyl-2-pyridinyl)piperazine ≥95%

Supplier: Matrix Scientific

072373-1G 072373-5G 072373-10G
102524-502EA 346.15 USD
102524-502 102524-504 102524-506
1-(5-Methyl-2-pyridinyl)piperazine ≥95%
1-(5-Methyl-2-pyridinyl)piperazine

Hazard: Irritant

Formula: C₁₀H₁₅N₃
MW: 177.25 g/mol
MDL Number: MFCD08061050
CAS Number: 104395-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR