(S)-α,α-Diphenyl-3-pyrrolidineacetamide ≥95%

Supplier: Matrix Scientific

072372-250MG 072372-1G 072372-5G
102524-496EA 498.46 USD
102524-496 102524-498 102524-500
(S)-α,α-Diphenyl-3-pyrrolidineacetamide ≥95%
(S)-alpha,alpha-Diphenyl-3-pyrrolidineacetamide

Hazard: Irritant

Formula: C₁₈H₂₀N₂O
CAS Number: 133099-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR