N-(2',6'-Dimethylphenyl)-2-piperidinecarboxamide ≥95%

Supplier: Matrix Scientific

Synonyms: Desbutylbupivacaine, N-Desbutyl bupivacaine, N-(2,6-Dimethylphenyl)piperidine-2-carboxamide

072370-1G 072370-5G
102524-486EA 710.77 USD
102524-486 102524-488
N-(2',6'-Dimethylphenyl)-2-piperidinecarboxamide ≥95%
N-(2',6'-Dimethylphenyl)-2-piperidinecarboxamide

Hazard: Irritant

Formula: C₁₄H₂₀N₂O
MW: 232.32 g/mol
MDL Number: MFCD01701244
CAS Number: 15883-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR