3-Bromo-5-nitropyridine ≥95%
Supplier: Matrix Scientific
072369-1G
072369-5G
102524-482EA
235.38
USD
102524-482
102524-484
3-Bromo-5-nitropyridine ≥95%
3-Bromo-5-nitropyridine
Hazard: Irritant
Formula:
C₅H₃BrN₂O₂ MW: 203 |
MDL Number:
MFCD04114216 CAS Number: 15862-30-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...