3-Amino-2,6-dimethoxypyridine ≥95%

Supplier: Matrix Scientific

072367-1G 072367-5G
102524-472EA 710.77 USD
102524-472 102524-474
3-Amino-2,6-dimethoxypyridine ≥95%
3-Amino-2,6-dimethoxypyridine

Hazard: Irritant

Formula: C7H10N2O2
MW: 154.1665 g/mol
MDL Number: MFCD00832865
CAS Number: 28020-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR