(4-Bromothiazol-5-yl)methanol ≥95%
Supplier: Matrix Scientific
072366-250MG
072366-1G
072366-5G
102524-466EA
640
USD
102524-466
102524-468
102524-470
(4-Bromothiazol-5-yl)methanol ≥95%
(4-Bromothiazol-5-yl)methanol
Hazard: Irritant
Formula:
C₄H₄BrNOS MW: 194.0497 g/mol |
MDL Number:
MFCD09909724 CAS Number: 262444-15-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...