tert-Butyl-4-((methylamino)methyl)piperidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 4-((methylamino)methyl)piperidine-1-carboxylate
072365-1G
072365-5G
072365-10G
102524-460EA
90.77
USD
102524-460
102524-462
102524-464
tert-Butyl-4-((methylamino)methyl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-((methylamino)methyl)piperidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₂H₂₄N₂O₂ MW: 228.3300 g/mol |
MDL Number:
MFCD06656629 CAS Number: 138022-02-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...