6-Chloroquinazolin-4-amine ≥95%

Supplier: Matrix Scientific

072364-250MG 072364-1G 072364-5G
102524-454EA 355.38 USD
102524-454 102524-456 102524-458
6-Chloroquinazolin-4-amine ≥95%
6-Chloroquinazolin-4-amine

Hazard: Irritant

Formula: C₈H₆ClN₃
MW: 179.6063 g/mol
MDL Number: MFCD02676502
CAS Number: 19808-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR