3,5-Dibromo-1H-indazole ≥95%

Supplier: Matrix Scientific

072363-1G 072363-5G
102524-450EA 640 USD
102524-450 102524-452
3,5-Dibromo-1H-indazole ≥95%
3,5-Dibromo-1H-indazole

Hazard: Irritant

Formula: C₇H₄Br₂N₂
MDL Number: MFCD00159919
CAS Number: 40598-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR