1-(2-Aminophenylsulfonyl)pyrrolidine ≥95%

Supplier: Matrix Scientific

072361-1G 072361-5G
102524-440EA 533.85 USD
102524-440 102524-442
1-(2-Aminophenylsulfonyl)pyrrolidine ≥95%
1-(2-Aminophenylsulphonyl)pyrrolidine

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₂S
MDL Number: MFCD09733905
CAS Number: 163460-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR