2-(4-((2-Aminophenyl)sulfonyl)piperazin-1-yl)ethanol ≥95%

Supplier: Matrix Scientific

072359-250MG 072359-1G 072359-5G
102524-430EA 498.46 USD
102524-430 102524-432 102524-434
2-(4-((2-Aminophenyl)sulfonyl)piperazin-1-yl)ethanol ≥95%
2-(4-((2-Aminophenyl)sulfonyl)piperazin-1-yl)ethanol

Hazard: Irritant

Formula: C₁₂H₁₉N₃O₃S
MW: 285.3626 g/mol
MDL Number: MFCD09909717
CAS Number: 1154310-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR