2-Amino-N-methylbenzenesulfonamide ≥95%
Supplier: Matrix Scientific
072356-1G
072356-5G
102524-418EA
569.23
USD
102524-418
102524-420
2-Amino-N-methylbenzenesulfonamide ≥95%
2-Amino-N-methylbenzenesulphonamide
Hazard: Irritant
Formula:
C₇H₁₀N₂O₂S |
MDL Number:
MFCD04035388 CAS Number: 16288-77-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...