2-Amino-N,N-dimethylbenzenesulfonamide ≥95%

Supplier: Matrix Scientific

072355-1G 072355-5G
102524-414EA 463.08 USD
102524-414 102524-416
2-Amino-N,N-dimethylbenzenesulfonamide ≥95%
2-Amino-N,N-dimethylbenzenesulphonamide

Hazard: Irritant

Formula: C₈H₁₂N₂O₂S
MDL Number: MFCD04035363
CAS Number: 54468-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR