tert-Butyl-5-chloro-6-(trifluoromethyl)-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

072353-250MG 072353-1G
102524-404EA 640 USD
102524-404 102524-406
tert-Butyl-5-chloro-6-(trifluoromethyl)-1H-indole-1-carboxylate ≥95%
tert-Butyl-5-chloro-6-(trifluoromethyl)-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₃ClF₃NO₂
CAS Number: 1209101-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR