(1-(tert-Butoxycarbonyl)-5,6-dichloro-1H-indol-2-yl)boronic acid ≥95%

Supplier: Matrix Scientific

072352-250MG 072352-1G
102524-400EA 924.62 USD
102524-400 102524-402
(1-(tert-Butoxycarbonyl)-5,6-dichloro-1H-indol-2-yl)boronic acid ≥95%
(1-(tert-Butoxycarbonyl)-5,6-dichloro-1H-indol-2-yl)boronic acid

Hazard: Irritant

Formula: C₁₃H₁₄BCl₂NO₄
CAS Number: 1310384-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR