1-Boc-5,6-dichloro-1H-indole ≥95%

Supplier: Matrix Scientific

072351-250MG 072351-1G 072351-5G
102524-394EA 355.38 USD
102524-394 102524-396 102524-398
1-Boc-5,6-dichloro-1H-indole ≥95%
1-Boc-5,6-dichloro-1H-indole

Hazard: Irritant

Formula: C₁₃H₁₃Cl₂NO₂
CAS Number: 1209183-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR