6-Bromothiazolo[4,5-b]pyrazin-2-amine ≥95%

Supplier: Matrix Scientific

072350-250MG 072350-1G
102524-390EA 640 USD
102524-390 102524-392
6-Bromothiazolo[4,5-b]pyrazin-2-amine ≥95%
6-Bromothiazolo[4,5-b]pyrazin-2-amine

Hazard: Irritant

Formula: C₅H₃BrN₄S
MW: 231.0731 g/mol
MDL Number: MFCD09909710
CAS Number: 112342-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR