5-Iodopyrimidine ≥95%

Supplier: Matrix Scientific

072349-1G 072349-5G
102524-386EA 500 USD
102524-386 102524-388
5-Iodopyrimidine ≥95%
5-Iodopyrimidine

Hazard: Irritant

Formula: C4H3IN2
MDL Number: MFCD00233961
CAS Number: 31462-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR