5-Bromo-2-chloro-7H-pyrrolo[2,3-d]pyrimidine ≥95%
Supplier: Matrix Scientific
072348-250MG
072348-1G
102524-380EA
883.08
USD
102524-380
102524-382
5-Bromo-2-chloro-7H-pyrrolo[2,3-d]pyrimidine ≥95%
5-Bromo-2-chloro-7H-pyrrolo[2,3-d]pyrimidine
Hazard: Irritant
Formula:
C₆H₃BrClN₃ MW: 232.4700 g/mol |
MDL Number:
MFCD09909707 CAS Number: 1060816-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...