7-Bromoquinazolin-4(3H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Bromo-3,4-dihydroquinazolin-4-one

072347-1G 072347-5G
102524-376EA 463.08 USD
102524-376 102524-378
7-Bromoquinazolin-4(3H)-one ≥95%
7-Bromoquinazolin-4(3H)-one

Hazard: Irritant

Formula: C₈H₅BrN₂O
MW: 225.0400 g/mol
MDL Number: MFCD07838323
CAS Number: 194851-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR