Ethyl-2-(5-bromopyrimidin-2-yl)acetate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(5-bromopyrimidin-2-yl)acetate

072345-250MG 072345-1G 072345-5G
102524-364EA 301 USD
102524-364 102524-366 102524-368
Ethyl-2-(5-bromopyrimidin-2-yl)acetate ≥95%
Ethyl-2-(5-bromopyrimidin-2-yl)acetate

Hazard: Irritant

Formula: C₈H₉BrN₂O₂
MW: 245.0733 g/mol
MDL Number: MFCD09909703
CAS Number: 1134327-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR