Ethyl-2-(pyrimidin-2-yl)acetate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(pyrimidin-2-yl)acetate, 2-Pyrimidineacetic acid ethyl ester

072344-250MG 072344-1G 072344-5G
102524-358EA 427.69 USD
102524-358 102524-360 102524-362
Ethyl-2-(pyrimidin-2-yl)acetate ≥95%
Ethyl-2-(pyrimidin-2-yl)acetate

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
MW: 166.1800 g/mol
MDL Number: MFCD09953517
CAS Number: 63155-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR