2-Chloro-5-iodopyrazine ≥95%

Supplier: Matrix Scientific

072342-250MG 072342-1G
102524-350EA 1066.15 USD
102524-350 102524-352
2-Chloro-5-iodopyrazine ≥95%
2-Chloro-5-iodopyrazine

Hazard: Irritant

Formula: C₄H₂ClIN₂
MW: 240.4300 g/mol
MDL Number: MFCD09909701
CAS Number: 1057216-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR