6-Bromo-4-chloro-2-methylquinazoline ≥95%

Supplier: Matrix Scientific

072339-250MG 072339-1G 072339-5G
102524-332EA 355.38 USD
102524-332 102524-334 102524-336
6-Bromo-4-chloro-2-methylquinazoline ≥95%
6-Bromo-4-chloro-2-methylquinazoline

Hazard: Irritant

Formula: C₉H₆BrClN₂
MW: 257.5143 g/mol
MDL Number: MFCD06657210
CAS Number: 351426-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR