6-Bromo-2-methylquinazolin-4(3H)-one ≥95%

Supplier: Matrix Scientific

072338-1G 072338-5G 072338-10G
102524-326EA 533.85 USD
102524-326 102524-328 102524-330
6-Bromo-2-methylquinazolin-4(3H)-one ≥95%
6-Bromo-2-methylquinazolin-4(3H)-one

Hazard: Irritant

Formula: C₉H₇BrN₂O
MDL Number: MFCD00548197
CAS Number: 5426-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR