6-Chloro-2-methylquinazolin-4(3H)-one ≥95%

Supplier: Matrix Scientific

072337-1G 072337-5G 072337-10G
102524-320EA 533.85 USD
102524-320 102524-322 102524-324
6-Chloro-2-methylquinazolin-4(3H)-one ≥95%
6-Chloro-2-methylquinazolin-4(3H)-one

Hazard: Irritant

Formula: C₉H₇ClN₂O
Melting Pt: 287 °C
MDL Number: MFCD00195330
CAS Number: 7142-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR