2-Amino-3-iodopyrazine ≥95%
Supplier: Matrix Scientific
072335-250MG
072335-1G
102524-310EA
924.62
USD
102524-310
102524-312
2-Amino-3-iodopyrazine ≥95%
2-Amino-3-iodopyrazine
Hazard: Irritant
Formula:
C₄H₄IN₃ |
CAS Number:
344329-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...