2-Fluoro-6-iodobenzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Fluoro-2-Iodobenzaldehyde

072333-1G 072333-5G 072333-10G
102524-300EA 123.08 USD
102524-300 102524-302 102524-304
2-Fluoro-6-iodobenzaldehyde ≥95%
2-Fluoro-6-iodobenzaldehyde

Hazard: Irritant

Formula: C₇H₄FIO
MW: 250.01 g/mol
MDL Number: MFCD07371506
CAS Number: 146137-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR